Thermo Scientific Chemicals

3-Phenylpyridine, 95%, Thermo Scientific Chemicals

Catalog number: B23930.06
5 g, Each
Thermo Scientific Chemicals

3-Phenylpyridine, 95%, Thermo Scientific Chemicals

Catalog number: B23930.06
5 g, Each
Quantity
Catalog number: B23930.06
also known as B23930-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1008-88-4
IUPAC Name
3-phenylpyridine
Molecular Formula
C11H9N
InChI Key
HJKGBRPNSJADMB-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=CC=CN=C1
Assay (GC)
≥94.0%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.6135-1.6195 @ 20?C
Form
Liquid

Description

3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text