Thermo Scientific Chemicals

o-Benzoic sulfimide, 98+%, Thermo Scientific Chemicals

Catalog number: B23938.0E
2500 g, Each
Thermo Scientific Chemicals

o-Benzoic sulfimide, 98+%, Thermo Scientific Chemicals

Catalog number: B23938.0E
2500 g, Each
Quantity
Catalog number: B23938.0E
also known as B23938-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81-07-2
IUPAC Name
2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
Molecular Formula
C7H5NO3S
InChI Key
CVHZOJJKTDOEJC-UHFFFAOYSA-N
SMILES
O=C1NS(=O)(=O)C2=CC=CC=C12
Form
Powder, crystals or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (HPLC)
≥98.0%
Melting Point (clear melt)
223-232?C

Description

o-Benzoic sulfimide is used as a pharmaceutic flavor aid. It is used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Benzoic sulfimide is used as a pharmaceutic flavor aid. It is used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame.

Solubility
Soluble in water (3.3 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text