Thermo Scientific Chemicals

6-Methoxy-2-naphthaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: B23943.06
5 g, Each
Thermo Scientific Chemicals

6-Methoxy-2-naphthaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: B23943.06
5 g, Each
Quantity
Catalog number: B23943.06
also known as B23943-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3453-33-6
Form
Crystals or powder or crystalline powder
Free acid (titration)
≤0.5%
Appearance (Color)
White to yellow to orange
Assay (GC)
≥98.5%
Melting Point (clear melt)
78.5-84.5?C

Description

6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text