6-Methoxy-2-naphthaldehyde, 99%
6-Methoxy-2-naphthaldehyde, 99%
6-Methoxy-2-naphthaldehyde, 99%
6-Methoxy-2-naphthaldehyde, 99%
Thermo Scientific Chemicals

6-Methoxy-2-naphthaldehyde, 99%

CAS: 3453-33-6 | C12H10O2 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B23943.14
also known as B23943-14
Price (USD)
162.00
Each
Estimated availability date 25-Jul-2025
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
162.00
Each
Add to cart
Chemical Identifiers
CAS3453-33-6
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤0.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White to yellow to orange
Assay (GC)≥98.5%
Melting Point (clear melt)78.5-84.5?C
Ask our AI about this Product
6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Alexander C. Goudie.; Laramie M. Gaster.; Antony W. Lake.; Carl J. Rose.; Patricia C. Freeman.; Bryn O. Hughes.; David Miller. 4-(6-Methoxy-2-naphthyl)butan-2-one and related analogs, a novel structural class of anti-inflammatory compounds. J. Med. Chem. 1978, 28 (12), 1260-1264.
  2. Jerry P. Jasinsk.; Ray J. Butcher.; Anil N. Mayekar.; H. S. Yathirajan.; B. Narayana. Structures of Three Chalcones Derived from 6-Methoxy-2-naphthaldehyde. Journal of Chemical Crystallography. 2009, 39 (3), 157-162.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
10231673Certificate of AnalysisApr 04, 2024B23943.06, B23943.14
10190106Certificate of AnalysisNov 23, 2021B23943.06, B23943.14
10231673Certificate of AnalysisNov 23, 2021B23943.06, B23943.14
10100206Certificate of AnalysisNov 23, 2021B23943.06, B23943.14
10049474Certificate of AnalysisNov 23, 2021B23943.06, B23943.14
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection