Thermo Scientific Chemicals

meso-Hydrobenzoin, 95%, Thermo Scientific Chemicals

Catalog number: B23946.14
25 g, Each
Thermo Scientific Chemicals

meso-Hydrobenzoin, 95%, Thermo Scientific Chemicals

Catalog number: B23946.14
25 g, Each
Quantity
Catalog number: B23946.14
also known as B23946-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
579-43-1
IUPAC Name
1,2-diphenylethane-1,2-diol
Molecular Formula
C14H14O2
InChI Key
IHPDTPWNFBQHEB-UHFFFAOYNA-N
SMILES
OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystalline and/or lumpy powder
Melting Point (clear melt)
134-140?C
Appearance (Color)
White
Assay (GC)
≥94.0% (sum of isomers)

Description

meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

Solubility
Soluble in hot alcohol or chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text