Thermo Scientific Chemicals

1-Iodo-4-nitrobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: B23950.22
100 g, Each
Thermo Scientific Chemicals

1-Iodo-4-nitrobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: B23950.22
100 g, Each
Quantity
Catalog number: B23950.22
also known as B23950-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
636-98-6
IUPAC Name
1-iodo-4-nitrobenzene
Molecular Formula
C6H4INO2
InChI Key
SCCCFNJTCDSLCY-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(I)C=C1
Assay (GC)
≥98.0%
Form
Powder
Melting Point (clear melt)
172-182?C
Appearance (Color)
Cream or pale yellow to brown
Identification (FTIR)
Conforms

Description

1-Iodo-4-nitrobenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-4-nitrobenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text