Thermo Scientific Chemicals

2-Hydroxy-1-naphthaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B23951.14
25 g, Each
Thermo Scientific Chemicals

2-Hydroxy-1-naphthaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B23951.14
25 g, Each
Quantity
Catalog number: B23951.14
also known as B23951-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
708-06-5
IUPAC Name
2-hydroxynaphthalene-1-carbaldehyde
Molecular Formula
C11H8O2
InChI Key
NTCCNERMXRIPTR-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CC=CC2=C1C=O
Appearance (Color)
White to yellow to pale brown
Melting Point (clear melt)
77-86?C
Form
Powder
Assay (HPLC)
≥97.5%
Assay (GC)
≥97.5%

Description

2-Hydroxy-1-naphthaldehyde is used in Merrified-type synthesis to deterime free amino acid groups in polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-1-naphthaldehyde is used in Merrified-type synthesis to deterime free amino acid groups in polymers.

Solubility
Soluble in water (40 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Store under dry inert gas. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text