Thermo Scientific Chemicals

Iron(II) fumarate, 94%, Thermo Scientific Chemicals

Catalog number: B23956.0B
1000 g, Each
Thermo Scientific Chemicals

Iron(II) fumarate, 94%, Thermo Scientific Chemicals

Catalog number: B23956.0B
1000 g, Each
Quantity
Catalog number: B23956.0B
also known as B23956-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-01-5
IUPAC Name
λ²-iron(2+) (2Z)-but-2-enedioate
Molecular Formula
C4H2FeO4
InChI Key
PMVSDNDAUGGCCE-ODZAUARKSA-L
SMILES
[Fe++].[O-]C(=O)C=C/C([O-])=O
Form
powder
Loss on Drying
< 1.5%
Appearance (Color)
Red to Brown
Impurity content
< 2.0% Iron(III)
Assay (unspecified)
> 92.5% (dry wt. basis)

Description

Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.

Solubility
Soluble in water (0.14 g/100 ml at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text