Iron(II) fumarate, 94%, Thermo Scientific Chemicals
Iron(II) fumarate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(II) fumarate, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B23956.30
also known as B23956-30
Price (USD)/ Each
44.65
Online exclusive
49.80 
Save 5.15 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
44.65
Online exclusive
49.80 
Save 5.15 (10%)
Add to cart
Iron(II) fumarate, 94%, Thermo Scientific Chemicals
Catalog numberB23956.30
Price (USD)/ Each
44.65
Online exclusive
49.80 
Save 5.15 (10%)
-
Add to cart
Chemical Identifiers
CAS141-01-5
IUPAC Nameλ²-iron(2+) (2Z)-but-2-enedioate
Molecular FormulaC4H2FeO4
InChI KeyPMVSDNDAUGGCCE-ODZAUARKSA-L
SMILES[Fe++].[O-]C(=O)C=C/C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Loss on Drying< 1.5%
Appearance (Color)Red to Brown
Impurity content< 2.0% Iron(III)
Assay (unspecified)> 92.5% (dry wt. basis)
Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.

Solubility
Soluble in water (0.14 g/100 ml at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Stanley H. Zlotkin.; Anna L. Christofides.; S. M. Ziauddin Hyder.; Claudia S. Schauer.; Mélody C. Tondeur.; Waseem Sharieff. Controlling iron deficiency anemia through the use of home-fortified complementary foods. The Indian Journal of Pediatrics. 2004, 78 (11), 1015-1019.
  2. Aparna Sil.; Vijaykumar S. Ijeri.; Ashwini K. Srivastava. Coated-wire iron(III) ion-selective electrode based on iron complex of 1,4,8,11-tetraazacyclotetradecane. Sensors and Actuators B: Chemical. 2005, 106 (2), 648-653.