Thermo Scientific™

2-(Bromoacetyl)naphthalene, 98%, Thermo Scientific Chemicals

Catalog number: B23960.18
50 g, Each
Thermo Scientific™

2-(Bromoacetyl)naphthalene, 98%, Thermo Scientific Chemicals

Catalog number: B23960.18
50 g, Each
Quantity
Catalog number: B23960.18
also known as B23960-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
613-54-7
IUPAC Name
2-bromo-1-(naphthalen-2-yl)ethan-1-one
Molecular Formula
C12H9BrO
InChI Key
YHXHHGDUANVQHE-UHFFFAOYSA-N
SMILES
BrCC(=O)C1=CC=C2C=CC=CC2=C1
Appearance (Color)
Pale pink to pink to purple to brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
79.5-85.5?C
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text