Thermo Scientific Chemicals

2-Bromophenylhydrazine hydrochloride, 94%, Thermo Scientific Chemicals

Catalog number: B23966.22
100 g, Each
Thermo Scientific Chemicals

2-Bromophenylhydrazine hydrochloride, 94%, Thermo Scientific Chemicals

Catalog number: B23966.22
100 g, Each
Quantity
Catalog number: B23966.22
also known as B23966-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50709-33-6
IUPAC Name
hydrogen (2-bromophenyl)hydrazine chloride
Molecular Formula
C6H8BrClN2
InChI Key
PHCYUJRYSFMJMG-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=CC=C1Br
Form
Powder or crystalline powder or lumps
Appearance (Color)
White to cream to pale brown
Proton NMR
Conforms to structure
Assay (Titration ex Chloride)
≤92.0 to ≤108.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text