Thermo Scientific™

Benzenesulfonyl hydrazide, 98%, Thermo Scientific Chemicals

Catalog number: B23971.36
500 g, Each
Thermo Scientific™

Benzenesulfonyl hydrazide, 98%, Thermo Scientific Chemicals

Catalog number: B23971.36
500 g, Each
Quantity
Catalog number: B23971.36
also known as B23971-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-17-1
IUPAC Name
benzenesulfonohydrazide
Molecular Formula
C6H8N2O2S
InChI Key
VJRITMATACIYAF-UHFFFAOYSA-N
SMILES
NNS(=O)(=O)C1=CC=CC=C1
Assay (unspecified)
≥97.5% (based on carbon EA)
Form
Powder or crystals
Elemental Analysis
Nitrogen N: 15.86-16.68% (theory 16.27%)
Appearance (Color)
White to pale cream
Elemental Analysis
Carbon C: 40.80-42.90% (theory 41.85%)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text