Furfurylamine, 99%, Thermo Scientific Chemicals
Furfurylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Furfurylamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number B23975.36
also known as B23975-36
Price (USD)/ Each
98.65
Online exclusive
109.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
98.65
Online exclusive
109.00 
Save 10.35 (10%)
Add to cart
Furfurylamine, 99%, Thermo Scientific Chemicals
Catalog numberB23975.36
Price (USD)/ Each
98.65
Online exclusive
109.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS617-89-0
IUPAC Name1-(furan-2-yl)methanamine
Molecular FormulaC5H7NO
InChI KeyDDRPCXLAQZKBJP-UHFFFAOYSA-N
SMILESNCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to yellow
Formliquid
Assay (GC)> 98.5%
Refractive Index1.4885 - 1.4915 @20?C
Furfurylamine is used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics, antihypertensive, and antiseptic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfurylamine is used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics, antihypertensive, and antiseptic agents.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are strong oxidizing agents, air, acids.
RUO – Research Use Only

General References:

  1. Kuo Yueh-Hsiung;Shih Kae-Shyang. A new method for preparation of 3-hydroxypyridines from furfurylamines by photooxygenation. Pharmaceutical Bulletin. 1991, 39 (1), 181-183.
  2. Rajappa Murali; H Surya Prakash Rao; Hans W Scheeren. Intra-molecular Diels-Alder reactions of citraconamic acids from furfurylamines and citraconic anhydride: effects of substitution in the furan ring on regioselectivity. Tetrahedron. 2001, 57 (15), 3165-3174.