Thermo Scientific Chemicals

Ethyl 2-pyridineacetate, 98%, Thermo Scientific Chemicals

Catalog number: B23988.14
25 g, Each
Thermo Scientific Chemicals

Ethyl 2-pyridineacetate, 98%, Thermo Scientific Chemicals

Catalog number: B23988.14
25 g, Each
Quantity
Catalog number: B23988.14
also known as B23988-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2739-98-2
IUPAC Name
ethyl 2-(pyridin-2-yl)acetate
Molecular Formula
C9H11NO2
InChI Key
IUDKTVXSXWAKJO-UHFFFAOYSA-N
SMILES
CCOC(=O)CC1=CC=CC=N1
Assay (GC)
≥97.5%
Appearance (Color)
Clear pale yellow to yellow
Refractive Index
1.4955-1.4995 @ 20?C
Form
Liquid

Description

Ethyl 2-pyridineacetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-pyridineacetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text