N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents and peroxide.
RUO – Research Use Only
General References:
- Chen, J.; Wang, Y.; Huang, Y.; Xu, K.; Li, N.; Wen, Q.; Zhou, Y. Magnetic multiwall carbon nanotubes modified with dual hydroxy functional ionic liquid for the solid-phase extraction of protein. Analyst 2015, 140 (10), 3474-3483.
- Wunderlich, C. H.; Juen, M. A.; LeBlanc, R. M.; Longhini, A. P.; Dayie, T. K.; Kreutz, C. Chapter Eighteen - Stable Isotope-Labeled RNA Phosphoramidites to Facilitate Dynamics by NMR. Methods Enzymol. 2015, 565, 461-494.