Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acids.
RUO – Research Use Only
Isao Mochida.; Seiichiro Eguchi.; Motohiro Hironaka.; Shin-ichi Nagao.; Kinya Sakanishi.; D. Duayne Whitehurst. The effects of seeding in the synthesis of zeolite MCM-22 in the presence of hexamethyleneimine. Zeolites. 1997, 18 (2-3), 142-151.
Douglas X. West.; Deborah S. Galloway. Transition metal ion complexes of thiosemicarbazones derived from 2-acetylpyridine. Part 3. The 3-hexamethyleneimine derivative. Transition Metal Chemistry. 1988, 13 (6), 410-414.