Thermo Scientific Chemicals

Hexamethyleneimine, 98+%, Thermo Scientific Chemicals

Catalog number: B24007.AP
500 mL, Each
Thermo Scientific Chemicals

Hexamethyleneimine, 98+%, Thermo Scientific Chemicals

Catalog number: B24007.AP
500 mL, Each
Quantity
Catalog number: B24007.AP
also known as B24007-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-49-9
IUPAC Name
azepane
Molecular Formula
C6H13N
InChI Key
ZSIQJIWKELUFRJ-UHFFFAOYSA-N
SMILES
C1CCCNCC1
Form
Liquid
Refractive Index
1.4645-1.4685 @ 20?C
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless to pale yellow

Description

Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text