Thermo Scientific Chemicals

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%, Thermo Scientific Chemicals

Catalog number: B24008.36
500 g, Each
Thermo Scientific Chemicals

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%, Thermo Scientific Chemicals

Catalog number: B24008.36
500 g, Each
Quantity
Catalog number: B24008.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6289-46-9
IUPAC Name
1,4-dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
Molecular Formula
C10H12O6
InChI Key
MHKKFFHWMKEBDW-UHFFFAOYNA-N
SMILES
COC(=O)C1CC(=O)C(CC1=O)C(=O)OC
Assay (HPLC)
≥99.0%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream to pale yellow or yellow-green

Description

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate is used in the production of pigments such as pigment red 122 and pigment violet 19. It also can react with 2,4-diamino-phenol to get 2,5-bis-(3-amino-4-hydroxy-phenylamino)-cyclohexa-1,4-diene-1,4-dicarboxylic acid dimethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate is used in the production of pigments such as pigment red 122 and pigment violet 19. It also can react with 2,4-diamino-phenol to get 2,5-bis-(3-amino-4-hydroxy-phenylamino)-cyclohexa-1,4-diene-1,4-dicarboxylic acid dimethyl ester.

Solubility
Soluble in water 0.0016 g/L.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text