Sodium 2,4-pentanedionate hydrate, 94%, Thermo Scientific Chemicals
Sodium 2,4-pentanedionate hydrate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium 2,4-pentanedionate hydrate, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number B24009.30
also known as B24009-30
Price (USD)/ Each
209.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
209.00
Add to cart
Sodium 2,4-pentanedionate hydrate, 94%, Thermo Scientific Chemicals
Catalog numberB24009.30
Price (USD)/ Each
209.00
-
Add to cart
Chemical Identifiers
CAS86891-03-4
IUPAC Namesodium 4-oxopent-2-en-2-olate
Molecular FormulaC5H7NaO2
InChI KeyAIWZOHBYSFSQGV-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
CommentMay contain small amounts of sodium hydroxide
Assay (Non-aqueous acid-base Titration)≥92.0 to ≤108.0% (dry weight basis)
Water Content (Karl Fischer Titration)≤26.95%
FormPowder
Sodium 2,4-pentanedionate hydrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 2,4-pentanedionate hydrate is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are strong oxidizing agents, water/ moisture, acids.
RUO – Research Use Only