N,N-Diethylacetoacetamide, 97%, Thermo Scientific Chemicals
N,N-Diethylacetoacetamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Diethylacetoacetamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number B24019.30
also known as B24019-30
Price (USD)/ Each
54.60
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
54.60
Add to cart
N,N-Diethylacetoacetamide, 97%, Thermo Scientific Chemicals
Catalog numberB24019.30
Price (USD)/ Each
54.60
-
Add to cart
Chemical Identifiers
CAS2235-46-3
IUPAC NameN,N-diethyl-3-oxobutanamide
Molecular FormulaC8H15NO2
InChI KeyNTMXFHGYWJIAAE-UHFFFAOYSA-N
SMILESCCN(CC)C(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4695-1.4745 @ 20?C
Appearance (Color)Clear yellow
Assay (GC)≥96.0%
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only