Thermo Scientific Chemicals

Ethyl 3-ethoxypropionate, 99+%, Thermo Scientific Chemicals

Catalog number: B24024.AU
1000 mL, Each
Thermo Scientific Chemicals

Ethyl 3-ethoxypropionate, 99+%, Thermo Scientific Chemicals

Catalog number: B24024.AU
1000 mL, Each
Quantity
Catalog number: B24024.AU
also known as B24024-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
763-69-9
IUPAC Name
ethyl 3-ethoxypropanoate
Molecular Formula
C7H14O3
InChI Key
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
SMILES
CCOCCC(=O)OCC
Form
Liquid
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4055-1.4075 @ 20?C
Assay (GC)
≥99.0%

Description

Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.

Solubility
Miscible with most organic solvents. Slightly miscible with cold water.

Notes
Incompatible with strong oxidizing agents, metals and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text