Thermo Scientific Chemicals

Hydroquinone bis(2-hydroxyethyl) ether, 95%, Thermo Scientific Chemicals

Catalog number: B24037.36
500 g, Each
Thermo Scientific Chemicals

Hydroquinone bis(2-hydroxyethyl) ether, 95%, Thermo Scientific Chemicals

Catalog number: B24037.36
500 g, Each
Quantity
Catalog number: B24037.36
also known as B24037-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-38-1
IUPAC Name
2-[4-(2-hydroxyethoxy)phenoxy]ethan-1-ol
Molecular Formula
C10H14O4
InChI Key
WTPYFJNYAMXZJG-UHFFFAOYSA-N
SMILES
OCCOC1=CC=C(OCCO)C=C1
Appearance (Color)
White to cream to pale brown
Melting Point (clear melt)
96.0-108.0?C
Assay (GC)
≥94.0%
Form
Flakes

Description

Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. Protect from humidity and water. It is hygroscopic in nature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text