2,4,6-Trimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals
2,4,6-Trimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Trimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B24060.14
also known as B24060-14
Price (USD)/ Each
298.65
Online exclusive
332.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
298.65
Online exclusive
332.00 
Save 33.35 (10%)
Add to cart
2,4,6-Trimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberB24060.14
Price (USD)/ Each
298.65
Online exclusive
332.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5980-97-2
IUPAC Name(2,4,6-trimethylphenyl)boronic acid
Molecular FormulaC9H13BO2
InChI KeyBZXQRXJJJUZZAJ-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(B(O)O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)117.0-127.0?C
Appearance (Color)White
Assay (HPLC)≥96.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only