Thermo Scientific Chemicals

1,4-Benzenediboronic acid, 96%, Thermo Scientific Chemicals

Catalog number: B24064.14
25 g, Each
Thermo Scientific Chemicals

1,4-Benzenediboronic acid, 96%, Thermo Scientific Chemicals

Catalog number: B24064.14
25 g, Each
Quantity
Catalog number: B24064.14
also known as B24064-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4612-26-4
IUPAC Name
[4-(dihydroxyboranyl)phenyl]boronic acid
Molecular Formula
C6H8B2O4
InChI Key
BODYVHJTUHHINQ-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(C=C1)B(O)O
Form
Powder
Assay (HPLC)
≥95.0%
Melting Point
>300?C
Appearance (Color)
White to pale cream
Identification (FTIR)
Conforms

Description

It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.

Solubility
Soluble in water 2.5%.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text