2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.
Solubility
Soluble in water 60 g/L at 20°C.
Notes
Store away from oxidizing agents, bases.
RUO – Research Use Only
General References:
- Nasser A Hassan; Mohamed I Hegab; Aymn E Rashad; Afaf A Fahmy;Synthesis and antimicrobial activity of some cyclic and acyclic nucleosides of thieno[2,3-d]pyrimidines. Nucleosides, Nucleotides & Nucleic Acids. 2007, 26 (4), 379-390.
- E. A. C. Lucken. Chemical applications of nuclear quadrupole resonance spectroscopy. Part III. The inductive effect of substituents belonging to the first row of the periodic table. J. Chem. Soc. 1959, 2954-2960.