Thermo Scientific Chemicals

2-Chloroethyl methyl ether, 98%, Thermo Scientific Chemicals

Catalog number: B24081.36
500 g, Each
Thermo Scientific Chemicals

2-Chloroethyl methyl ether, 98%, Thermo Scientific Chemicals

Catalog number: B24081.36
500 g, Each
Quantity
Catalog number: B24081.36
also known as B24081-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
627-42-9
IUPAC Name
1-chloro-2-methoxyethane
Molecular Formula
C3H7ClO
InChI Key
XTIGGAHUZJWQMD-UHFFFAOYSA-N
SMILES
COCCCl
Form
Liquid
Refractive Index
1.4070-1.4110 @ 20?C
Appearance (Color)
Clear, colorless to yellow
Assay (GC)
>97.5%

Description

2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

Solubility
Soluble in water 60 g/L at 20°C.

Notes
Store away from oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text