Thermo Scientific Chemicals

2-Methoxypropene, 95%, stab. with ca 0.5% potassium carbonate, Thermo Scientific Chemicals

Catalog number: B24083.18
50 g, Each
Thermo Scientific Chemicals

2-Methoxypropene, 95%, stab. with ca 0.5% potassium carbonate, Thermo Scientific Chemicals

Catalog number: B24083.18
50 g, Each
Quantity
Catalog number: B24083.18
also known as B24083-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
116-11-0
IUPAC Name
2-methoxyprop-1-ene
Molecular Formula
C4H8O
InChI Key
YOWQWFMSQCOSBA-UHFFFAOYSA-N
SMILES
COC(C)=C
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%
Refractive Index
1.3805-1.3865 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool, dry place in tightly closed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text