Chemical Identifiers
CAS6424-15-3
IUPAC Namebis(2-hydroxypropane-1,2,3-tricarboxylic acid) tricobalt dihydrate
Molecular FormulaC12H20Co3O16
InChI KeySKMIIAIULKZNRQ-UHFFFAOYSA-N
SMILESO.O.[Co].[Co].[Co].OC(=O)CC(O)(CC(O)=O)C(O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O
View more