Thermo Scientific Chemicals

Methyl 4-formylbenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: B24091.22
100 g, Each
Thermo Scientific Chemicals

Methyl 4-formylbenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: B24091.22
100 g, Each
Quantity
Catalog number: B24091.22
also known as B24091-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1571-08-0
IUPAC Name
methyl 4-formylbenzoate
Molecular Formula
C9H8O3
InChI Key
FEIOASZZURHTHB-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(C=O)C=C1
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%
Free acid (titration)
≤1.5% (non-U.S. specification)
Appearance (Color)
White to cream to pale yellow
Melting Point (clear melt)
58.0-65.0°C (non-U.S. specification)

Description

Methyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text