4-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24117.03
also known as B24117-03
Price (USD)/ Each
79.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
79.00
Add to cart
4-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Catalog numberB24117.03
Price (USD)/ Each
79.00
-
Add to cart
Chemical Identifiers
CAS139911-29-8
IUPAC Name(4-fluoro-2-methylphenyl)boronic acid
Molecular FormulaC7H8BFO2
InChI KeyIQMLIVUHMSIOQP-UHFFFAOYSA-N
SMILESCC1=CC(F)=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only