Thermo Scientific Chemicals

2,3-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24125.14
25 g, Each
Thermo Scientific Chemicals

2,3-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24125.14
25 g, Each
Quantity
Catalog number: B24125.14
also known as B24125-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40972-86-9
IUPAC Name
(2,3-dimethoxyphenyl)boronic acid
Molecular Formula
C8H11BO4
InChI Key
VREWSCMOGIXMDQ-UHFFFAOYSA-N
SMILES
COC1=CC=CC(B(O)O)=C1OC
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.0%
Proton NMR
Conforms to structure
Assay (Aqueous acid-base Titration)
≥97.0%
Appearance (Color)
White to pale cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text