Thermo Scientific Chemicals

tert-Butylcyclohexane, 99+%, Thermo Scientific Chemicals

Catalog number: B24126.14
25 g, Each
Thermo Scientific Chemicals

tert-Butylcyclohexane, 99+%, Thermo Scientific Chemicals

Catalog number: B24126.14
25 g, Each
Quantity
Catalog number: B24126.14
also known as B24126-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3178-22-1
IUPAC Name
tert-butylcyclohexane
Molecular Formula
C10H20
InChI Key
XTVMZZBLCLWBPM-UHFFFAOYSA-N
SMILES
CC(C)(C)C1CCCCC1
Refractive Index
1.4460-1.4480 @ 20?C
Form
Liquid
Assay (GC)
≥99.0%
Appearance (Color)
Clear colorless

Description

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text