Thermo Scientific Chemicals

(1-Butyl)triethylammonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: B24128.22
100 g, Each
Thermo Scientific Chemicals

(1-Butyl)triethylammonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: B24128.22
100 g, Each
Quantity
Catalog number: B24128.22
also known as B24128-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13028-69-8
IUPAC Name
butyltriethylazanium
Molecular Formula
C10H24N
InChI Key
MPDDDPYHTMZBMG-UHFFFAOYSA-N
SMILES
CCCC[N+](CC)(CC)CC
Assay (Titration ex Bromide)
≥96.0 to ≤104.0%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤1.5%

Description

(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text