Thermo Scientific Chemicals

Methyl alpha-L-rhamnopyranoside, 98%, Thermo Scientific Chemicals

Catalog number: B24129.14
25 g, Each
Thermo Scientific Chemicals

Methyl alpha-L-rhamnopyranoside, 98%, Thermo Scientific Chemicals

Catalog number: B24129.14
25 g, Each
Quantity
Catalog number: B24129.14
also known as B24129-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14917-55-6
IUPAC Name
2-methoxy-6-methyloxane-3,4,5-triol
Molecular Formula
C7H14O5
InChI Key
OHWCAVRRXKJCRB-UHFFFAOYNA-N
SMILES
COC1OC(C)C(O)C(O)C1O
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥97.5%
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text