Thermo Scientific Chemicals

4-Methylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B24139.09
10 g, Each
Thermo Scientific Chemicals

4-Methylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B24139.09
10 g, Each
Quantity
Catalog number: B24139.09
also known as B24139-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
822-36-6
IUPAC Name
5-methyl-1H-imidazole
Molecular Formula
C4H6N2
InChI Key
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
SMILES
CC1=CN=CN1
Appearance (Color)
White to cream to yellow
Form
Crystalline powder or crystals or fused solid and/or chunks
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point
49-57°C

Description

4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text