4'-Bromo-4-chlorobutyrophenone is used as a pharmaceutical & syntheses material intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Bromo-4-chlorobutyrophenone is used as a pharmaceutical & syntheses material intermediates.
Solubility
Insoluble in water.
Notes
Storage temperature is 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
RUO – Research Use Only
D. Martin Davies; Michael E. Deary and David I. Wealleans. Stability constants of α-cyclodextrin complexes of para-substituted aromatic ketones in aqueous solution. J. Chem. Soc., Perkin Trans. 2. 1998, (1), 193-196.
Maris Vilums; Annelien J.M. Zweemer; Farhana Barmare; Anouk M.F. van der Gracht; Dave C.T. Bleeker; Zhiyi Yua, Henk de Vries; Raymond Gross; Jeremy Clemens; Paul Krenitsky; Johannes Brussee; Dean Stamos; John Saunders; Laura H. Heitman; Adriaan P. IJzerman. When structure-affinity relationships meet structure-kinetics relationships: 3-((Inden-1-yl)amino)-1-isopropyl-cyclopentane-1-carboxamides as CCR2 antagonists. European Journal of Medicinal Chemistry. 2015, 93, 121-134.