Thermo Scientific Chemicals

2-Naphthaleneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B24157.14
25 g, Each
Thermo Scientific Chemicals

2-Naphthaleneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B24157.14
25 g, Each
Quantity
Catalog number: B24157.14
also known as B24157-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32316-92-0
IUPAC Name
(naphthalen-2-yl)boronic acid
Molecular Formula
C10H9BO2
InChI Key
KPTRDYONBVUWPD-UHFFFAOYSA-N
SMILES
OB(O)C1=CC2=CC=CC=C2C=C1
Identification (FTIR)
Conforms
Proton NMR
Conforms to structure
Appearance (Color)
White to cream to pale brown
Form
Powder or crystalline powder and/or chunks
Assay (HPLC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text