Thermo Scientific Chemicals

2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: B24161.36
500 g, Each
Thermo Scientific Chemicals

2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: B24161.36
500 g, Each
Quantity
Catalog number: B24161.36
also known as B24161-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
550-99-2
IUPAC Name
hydrogen 2-[(naphthalen-1-yl)methyl]-4,5-dihydro-1H-imidazole chloride
Molecular Formula
C14H15ClN2
InChI Key
DJDFFEBSKJCGHC-UHFFFAOYSA-N
SMILES
[H+].[Cl-].C(C1=NCCN1)C1=C2C=CC=CC2=CC=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (HPLC)
≥98.5%
Identification (FTIR)
Conforms

Description

Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 170 g/L, alcohol, DMSO, ethanol.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text