Thermo Scientific Chemicals

Mucic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24170.22
100 g, Each
Thermo Scientific Chemicals

Mucic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24170.22
100 g, Each
Quantity
Catalog number: B24170.22
also known as B24170-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
526-99-8
IUPAC Name
2,3,4,5-tetrahydroxyhexanedioic acid
Molecular Formula
C6H10O8
InChI Key
DSLZVSRJTYRBFB-UHFFFAOYNA-N
SMILES
OC(C(O)C(O)C(O)=O)C(O)C(O)=O
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
Conforms to structure
Solution Test
4% wt (1N NaOH solution): Clear, colorless solution
Form
Crystalline powder
Sulfated Ash
≤0.1%

Description

Mucic acid is used to replace tartaric acid in self-rising flour or fizzies. It is also used as a precursor of adipic acid en route to nylon by a rhenium-catalyzed deoxydehydration reaction. It replaces potassium bitartrate in baking powder and also used for manufacture of granular effervescing salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mucic acid is used to replace tartaric acid in self-rising flour or fizzies. It is also used as a precursor of adipic acid en route to nylon by a rhenium-catalyzed deoxydehydration reaction. It replaces potassium bitartrate in baking powder and also used for manufacture of granular effervescing salts.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text