N-(n-Butyl)aniline, 99%, Thermo Scientific Chemicals
N-(n-Butyl)aniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(n-Butyl)aniline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number B24180.22
also known as B24180-22
Price (USD)/ Each
216.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
216.00
Add to cart
N-(n-Butyl)aniline, 99%, Thermo Scientific Chemicals
Catalog numberB24180.22
Price (USD)/ Each
216.00
-
Add to cart
Chemical Identifiers
CAS1126-78-9
IUPAC NameN-butylaniline
Molecular FormulaC10H15N
InChI KeyVSHTWPWTCXQLQN-UHFFFAOYSA-N
SMILESCCCCNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to yellow to orange to brown
FormLiquid
Refractive Index1.5320-1.5350 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only