Thermo Scientific Chemicals

Hexarhodium hexadecacarbonyl, 98%, Thermo Scientific Chemicals

Catalog number: B24185.01
0.25 g, Each
Thermo Scientific Chemicals

Hexarhodium hexadecacarbonyl, 98%, Thermo Scientific Chemicals

Catalog number: B24185.01
0.25 g, Each
Quantity
Catalog number: B24185.01
also known as B24185-01
Price (USD)
Have Questions?

Chemical Identifiers

CAS
28407-51-4
IUPAC Name
hexadecakis(methanidylidyneoxidanium) hexarhodium
Molecular Formula
C16O16Rh6
InChI Key
SZQABOJVTZVBHE-UHFFFAOYSA-N
SMILES
[Rh].[Rh].[Rh].[Rh].[Rh].[Rh].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Assay (unspecified)
≥97.5 to ≤102.5% (Based on elemental Rh)
Elemental Analysis
C: 17.58-18.49% (Theory: 18.03%)
Appearance (Color)
Black or dark blue
Elemental Analysis
Rh: 56.49-59.39% (Theory: 57.94%)
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text