Thermo Scientific Chemicals

Ethyl nonanoate, 97%, Thermo Scientific Chemicals

Catalog number: B24190.AP
500 mL, Each
Thermo Scientific Chemicals

Ethyl nonanoate, 97%, Thermo Scientific Chemicals

Catalog number: B24190.AP
500 mL, Each
Quantity
Catalog number: B24190.AP
also known as B24190-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-29-5
IUPAC Name
ethyl nonanoate
Molecular Formula
C11H22O2
InChI Key
BYEVBITUADOIGY-UHFFFAOYSA-N
SMILES
CCCCCCCCC(=O)OCC
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.4190-1.4240 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text