6-Methyluracil, 97%, Thermo Scientific Chemicals
6-Methyluracil, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methyluracil, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number B24191.22
also known as B24191-22
Price (USD)/ Each
28.50
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
28.50
Add to cart
6-Methyluracil, 97%, Thermo Scientific Chemicals
Catalog numberB24191.22
Price (USD)/ Each
28.50
-
Add to cart
Chemical Identifiers
CAS626-48-2
IUPAC Name6-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC5H6N2O2
InChI KeySHVCSCWHWMSGTE-UHFFFAOYSA-N
SMILESCC1=CC(=O)NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only