Thermo Scientific Chemicals

Phenolphthalein disulfate tripotassium salt hydrate, 98%

Catalog number: B24198.03
1 g, Each
Thermo Scientific Chemicals

Phenolphthalein disulfate tripotassium salt hydrate, 98%

Catalog number: B24198.03
1 g, Each
Quantity
Catalog number: B24198.03
also known as B24198-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62625-16-5
IUPAC Name
tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate
Molecular Formula
C20H13K3O11S2
InChI Key
PLHSVCWOEHYSHO-UHFFFAOYSA-K
SMILES
[K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O
Form
Powder or crystalline powder
Appearance (Color)
White
Identification (FTIR)
Conforms
Assay (HPLC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text