Thermo Scientific Chemicals

1-Iodoheptane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B24206.18
50 g, Each
Thermo Scientific Chemicals

1-Iodoheptane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B24206.18
50 g, Each
Quantity
Catalog number: B24206.18
also known as B24206-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4282-40-0
IUPAC Name
1-iodoheptane
Molecular Formula
C7H15I
InChI Key
LMHCYRULPLGEEZ-UHFFFAOYSA-N
SMILES
CCCCCCCI
Form
Liquid
Comment
Copper powder used as stabilizer
Refractive Index
1.4880-1.4920 @ 20?C
Appearance (Color)
Clear colorless to pale pink
Assay (GC)
>97.5%

Description

1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text