Thermo Scientific Chemicals

Methyl vanillate, 99%, Thermo Scientific Chemicals

Catalog number: B24209.06
5 g, Each
Thermo Scientific Chemicals

Methyl vanillate, 99%, Thermo Scientific Chemicals

Catalog number: B24209.06
5 g, Each
Quantity
Catalog number: B24209.06
also known as B24209-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3943-74-6
IUPAC Name
methyl 4-hydroxy-3-methoxybenzoate
Molecular Formula
C9H10O4
InChI Key
BVWTXUYLKBHMOX-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(O)C(OC)=C1
Appearance (Color)
White to cream to pale brown
Form
Crystals or crystalline powder or powder
Water Content (Karl Fischer Titration)
≤1.0%
Assay (GC)
≥98.5%
Melting Point (clear melt)
62-67°C

Description

Methyl vanillate is used to investigate the volatile compounds of heather honeys from Calluna vulgaris and Erica arborea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl vanillate is used to investigate the volatile compounds of heather honeys from Calluna vulgaris and Erica arborea.

Solubility
Soluble in chloroform and methanol.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text