Methyl vanillate is used to investigate the volatile compounds of heather honeys from Calluna vulgaris and Erica arborea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl vanillate is used to investigate the volatile compounds of heather honeys from Calluna vulgaris and Erica arborea.
Solubility
Soluble in chloroform and methanol.
Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
Tobias Voitl.; Philipp Rudolf von Rohr. Demonstration of a Process for the Conversion of Kraft Lignin into Vanillin and Methyl Vanillate by Acidic Oxidation in Aqueous Methanol. Ind. Eng. Chem. Res. 2007, 49 (2), 520-525.
C. Saiz-Jimenez.; J.W. de Leeuw. Pyrolysis-gas chromatography-mass spectrometry of isolated, synthetic and degraded lignins. Organic Geochemistry. 1984, 6 (21), 417-422.