Thermo Scientific Chemicals

Zinc formate, 98%, Thermo Scientific Chemicals

Catalog number: B24210.22
100 g, Each
Thermo Scientific Chemicals

Zinc formate, 98%, Thermo Scientific Chemicals

Catalog number: B24210.22
100 g, Each
Quantity
Catalog number: B24210.22
also known as B24210-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-41-5
IUPAC Name
zinc(2+) diformate
Molecular Formula
C2H2O4Zn
InChI Key
XOBMCBQSUCOAOC-UHFFFAOYSA-L
SMILES
[Zn++].[O-]C=O.[O-]C=O
Water Content (Karl Fischer Titration)
≤3%
Complexometric Titration
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White

Description

Zinc formate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc formate is used as a pharmaceutical intermediate.

Solubility
Soluble in water (52 g/L at 20°C).

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text