3,4-(Methylenedioxy)benzeneboronic acid, 98%, Thermo Scientific Chemicals
3,4-(Methylenedioxy)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-(Methylenedioxy)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B24217.06
also known as B24217-06
Price (USD)/ Each
140.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
140.00
Add to cart
3,4-(Methylenedioxy)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Catalog numberB24217.06
Price (USD)/ Each
140.00
-
Add to cart
Chemical Identifiers
CAS94839-07-3
IUPAC Name(2H-1,3-benzodioxol-5-yl)boronic acid
Molecular FormulaC7H7BO4
InChI KeyCMHPUBKZZPSUIQ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Aqueous acid-base Titration)≥97.5%
Appearance (Color)White to cream to yellow to pale brown
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only