Thermo Scientific Chemicals

2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B24232.06
5 g, Each
Thermo Scientific Chemicals

2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B24232.06
5 g, Each
Quantity
Catalog number: B24232.06
also known as B24232-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
656-42-8
IUPAC Name
2,2-difluoro-2H-1,3-benzodioxole-5-carbaldehyde
Molecular Formula
C8H4F2O3
InChI Key
GGERGLKEDUUSAP-UHFFFAOYSA-N
SMILES
FC1(F)OC2=CC=C(C=O)C=C2O1
Free acid (titration)
≤1.5%
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.4960-1.5010 @ 20?C

Description

2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde is used as primary and secondary intermediate.

Solubility
Sparingly soluble in water (0.31 g/L at 25°C).

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text